4-(2-Fluoroethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

426696-500MG 426696-5G 426696-25G
77483-860EA 179.22 USD
77483-860 77494-326 77503-998
4-(2-Fluoroethoxy)benzaldehyde ≥95%
4-(2-Fluoroethoxy)benzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
CAS Number: 2967-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR