1-(4-(cyclopentyloxy)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

425794-500MG 425794-5G 425794-25G
77483-856EA 202.91 USD
77483-856 77494-322 77503-994
1-(4-(cyclopentyloxy)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-(cyclopentyloxy)phenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₃H₁₃F₃O₂
MW: 258.2363 g/mol
Storage Temperature: Ambient
CAS Number: 1368690-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR