Ethyl 4-chloro-α-oxobenzo[b]thiophene-2-acetate ≥95%

Supplier: AOBChem USA

Warning

423912-500MG 423912-5G
77483-852EA 436.72 USD
77483-852 77494-318
Ethyl 4-chloro-α-oxobenzo[b]thiophene-2-acetate ≥95%
Ethyl 4-chloro-α-oxobenzo[b]thiophene-2-acetate

Precaución: For research use only

Formula: C₁₂H₉ClO₃S
MW: 268.72 g/mol
Storage Temperature: Ambient
CAS Number: 1897819-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR