1,5-Dibromo-3-fluoro-2,4-Dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

423565-500MG 423565-5G
77483-828EA 462.47 USD
77483-828 77494-294
1,5-Dibromo-3-fluoro-2,4-Dimethylbenzene ≥95%
1,5-Dibromo-3-fluoro-2,4-Dimethylbenzene

Precaución: For research use only

Formula: C₈H₇Br₂F
MW: 281.9475 g/mol
Storage Temperature: Ambient
CAS Number: 1781113-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR