4-Bromo-2,6-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

42318-500MG 42318-5G 42318-25G
77483-824EA 298.7 USD
77483-824 77494-290 77503-970
4-Bromo-2,6-dimethoxybenzoic acid ≥97%
4-Bromo-2,6-dimethoxybenzoic acid

Precaución: For research use only

Formula: C₉H₉BrO₄
Storage Temperature: Ambient
MDL Number: MFCD28132241
CAS Number: 61203-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR