1-(3-Chloropyridin-4-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

422682-500MG 422682-5G 422682-25G
77483-820EA 252.35 USD
77483-820 77494-286 77503-968
1-(3-Chloropyridin-4-yl)propan-1-ol ≥95%
1-(3-Chloropyridin-4-yl)propan-1-ol

Precaución: For research use only

Formula: C₈H₁₀ClNO
MW: 171.6241 g/mol
Storage Temperature: Ambient
CAS Number: 1600189-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR