1-(5-Bromothiophen-2-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

421287-5G 421287-25G
77494-262EA 1065.02 USD
77494-262 77503-948
1-(5-Bromothiophen-2-yl)propan-1-ol ≥95%
1-(5-Bromothiophen-2-yl)propan-1-ol

Precaución: For research use only

Formula: C₇H₉BrOS
CAS Number: 1216266-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR