2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2,3-Difluoro-4-(trifluoromethyl)phenyl)boronic acid, [2,3-Difluoro-4-(trifluoromethyl)phenyl]boronic acid

42042-5G 42042-25G
77494-246EA 230.72 USD
77494-246 77503-936
2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid ≥95%
2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid

Precaución: For research use only

Formula: C₇H₄BF₅O₂
Storage Temperature: Ambient
MDL Number: MFCD14892600
CAS Number: 2096332-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR