(3-Bromo-2,6-difluorophenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

415126-5G 415126-25G
77494-238EA 1484.23 USD
77494-238 77503-928
(3-Bromo-2,6-difluorophenyl)(phenyl)methanone ≥95%
(3-Bromo-2,6-difluorophenyl)(phenyl)methanone

Precaución: For research use only

Formula: C₁₃H₇BrF₂O
CAS Number: 1970303-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR