(5-Bromo-4-methylthiophen-2-yl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

414982-5G 414982-25G
77494-226EA 1409.04 USD
77494-226 77503-920
(5-Bromo-4-methylthiophen-2-yl)(cyclopropyl)methanone ≥95%
(5-Bromo-4-methylthiophen-2-yl)(cyclopropyl)methanone

Precaución: For research use only

Formula: C₉H₉BrOS
CAS Number: 1537122-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR