3,4-Dichloro-2-methylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3,4-Dichloro-2-methylphenyl)boronic acid

41333-1G 41333-500MG 41333-5G 41333-25G
77471-096EA 251.32 USD
77471-096 77483-752 77494-214 77503-910
3,4-Dichloro-2-methylphenylboronic acid ≥95%
3,4-Dichloro-2-methylphenylboronic acid

Precaución: For research use only

Formula: C₇H₇BCl₂O₂
MW: 204.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475873
CAS Number: 957035-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR