2-Chloro-5-ethoxyphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
41262-5G
41262-25G
77494-210EA
224.54
USD
77494-210
77503-906
2-Chloro-5-ethoxyphenylboronic acid ≥97%
2-Chloro-5-ethoxyphenylboronic acid
Precaución: For research use only
Formula:
C₈H₁₀BClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD08235081 CAS Number: 913835-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...