[4-(Allyloxy)phenyl]boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Allyloxyphenylboronic acid, (4-(Allyloxy)phenyl)boronic acid

39524-5G 39524-25G
77494-174EA 224.54 USD
77494-174 77503-874
[4-(Allyloxy)phenyl]boronic acid ≥95%
[4-(Allyloxy)phenyl]boronic acid

Precaución: For research use only

Formula: C₉H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD11103728
CAS Number: 1117776-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR