Methyl(3-propoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

394384-5G 394384-25G
77494-166EA 924.94 USD
77494-166 77503-866
Methyl(3-propoxyphenyl)sulfane ≥95%
Methyl(3-propoxyphenyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₄OS
CAS Number: 178987-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR