6-Bromo-2-fluoro-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

38623-5G 38623-25G
77494-106EA 677.74 USD
77494-106 77503-816
6-Bromo-2-fluoro-3-isopropoxybenzaldehyde ≥97%
6-Bromo-2-fluoro-3-isopropoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MDL Number: MFCD32707102
CAS Number: 2413441-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR