2-(3-Bromo-2,6-dimethoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

38270-5G 38270-25G
77494-086EA 1303.98 USD
77494-086 77503-802
2-(3-Bromo-2,6-dimethoxyphenyl)-1,3-dioxolane ≥95%
2-(3-Bromo-2,6-dimethoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃BrO₄
MDL Number: MFCD34564066
CAS Number: 2734772-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR