(4-(phenylethynyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

380856-5G 380856-25G
77494-054EA 1493.5 USD
77494-054 77503-770
(4-(phenylethynyl)phenyl)boronic acid ≥95%
(4-(phenylethynyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₁₄H₁₁BO₂
CAS Number: 702699-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR