1-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

380454-5G 380454-25G
77494-044EA 1484.23 USD
77494-044 77503-760
1-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)propan-1-one ≥95%
1-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)propan-1-one

Precaución: For research use only

Formula: C₁₆H₁₄ClFO₂
CAS Number: 3002463-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR