1-(4-Fluoro-2-isopropoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

380298-5G 380298-25G
77494-032EA 543.84 USD
77494-032 77503-750
1-(4-Fluoro-2-isopropoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Fluoro-2-isopropoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₁FO₂
CAS Number: 3002481-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR