2,2,2-trifluoro-1-(4-fluoro-2-isopropoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

380287-5G 380287-25G
77494-024EA 1484.23 USD
77494-024 77503-742
2,2,2-trifluoro-1-(4-fluoro-2-isopropoxyphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(4-fluoro-2-isopropoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₀F₄O₂
CAS Number: 3002449-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR