Cyclobutyl(4-fluoro-2-isopropoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

380281-5G 380281-25G
77494-020EA 1484.23 USD
77494-020 77503-738
Cyclobutyl(4-fluoro-2-isopropoxyphenyl)methanone ≥95%
Cyclobutyl(4-fluoro-2-isopropoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₄H₁₇FO₂
CAS Number: 3002553-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR