(2-Ethoxy-4-fluorophenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

380110-5G 380110-25G
77494-008EA 1484.23 USD
77494-008 77503-726
(2-Ethoxy-4-fluorophenyl)(isopropyl)sulfane ≥95%
(2-Ethoxy-4-fluorophenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₅FOS
CAS Number: 3002531-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR