Cyclopropyl(4-fluoro-2-propoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

380059-5G 380059-25G
77493-992EA 1484.23 USD
77493-992 77503-710
Cyclopropyl(4-fluoro-2-propoxyphenyl)methanol ≥95%
Cyclopropyl(4-fluoro-2-propoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₇FO₂
CAS Number: 3002473-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR