Ethyl(4-fluoro-2-propoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

380056-5G 380056-25G
77493-988EA 1484.23 USD
77493-988 77503-706
Ethyl(4-fluoro-2-propoxyphenyl)sulfane ≥95%
Ethyl(4-fluoro-2-propoxyphenyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₅FOS
CAS Number: 3002521-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR