1,2,4-tribromo-3-fluorobenzene ≥95%

Supplier: AOBChem USA

378800-5G 378800-25G
77493-944EA 1484.23 USD
77493-944 77503-668
1,2,4-tribromo-3-fluorobenzene ≥95%
1,2,4-tribromo-3-fluorobenzene

Precaución: For research use only

Formula: C₆H₂Br₃F
CAS Number: 811711-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR