1-(5-Bromo-2,3,4-trifluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

377980-5G 377980-25G
77493-920EA 1484.23 USD
77493-920 77503-648
1-(5-Bromo-2,3,4-trifluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(5-Bromo-2,3,4-trifluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₂BrF₃O
MW: 297.112 g/mol
CAS Number: 3002529-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR