1-(2,6-Dichloro-3-ethoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

375947-5G 375947-25G
77493-876EA 1484.23 USD
77493-876 77503-610
1-(2,6-Dichloro-3-ethoxyphenyl)-2,2-difluoroethanone ≥95%
1-(2,6-Dichloro-3-ethoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₈Cl₂F₂O₂
CAS Number: 3002521-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR