(4-Bromo-3-fluoro-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

37424-5G 37424-25G
77493-852EA 1509.98 USD
77493-852 77503-592
(4-Bromo-3-fluoro-2-methylphenyl)(methyl)sulfane ≥95%
(4-Bromo-3-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₈BrFS
MDL Number: MFCD34178500
CAS Number: 2643367-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR