α-(3-Phenoxy-1-propyn-1-yl)benzenemethanol ≥95%

Supplier: AOBChem USA

372347-5G 372347-25G
77493-840EA 811.64 USD
77493-840 77503-582
α-(3-Phenoxy-1-propyn-1-yl)benzenemethanol ≥95%
α-(3-Phenoxy-1-propyn-1-yl)benzenemethanol

Precaución: For research use only

Formula: C₁₆H₁₄O₂
CAS Number: 32833-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR