4-Bromo-1-fluoro-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

37212-5G 37212-25G
77493-824EA 494.4 USD
77493-824 77503-566
4-Bromo-1-fluoro-2-naphthaldehyde ≥97%
4-Bromo-1-fluoro-2-naphthaldehyde

Precaución: For research use only

Formula: C₁₁H₆BrFO
MDL Number: MFCD11100825
CAS Number: 842136-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR