(4-Bromo-2,3-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

37066-5G 37066-25G
77493-810EA 543.84 USD
77493-810 77503-554
(4-Bromo-2,3-dimethylphenyl)(methyl)sulfane ≥95%
(4-Bromo-2,3-dimethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁BrS
MDL Number: MFCD32707015
CAS Number: 250593-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR