1-(3-Bromo-2,6-dichlorophenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

369977-5G 369977-25G
77493-800EA 1484.23 USD
77493-800 77503-546
1-(3-Bromo-2,6-dichlorophenyl)prop-2-en-1-ol ≥95%
1-(3-Bromo-2,6-dichlorophenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₉H₇BrCl₂O
CAS Number: 3002462-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR