(3-Bromo-2,6-dichlorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

369973-5G 369973-25G
77493-796EA 1484.23 USD
77493-796 77503-542
(3-Bromo-2,6-dichlorophenyl)(phenyl)methanol ≥95%
(3-Bromo-2,6-dichlorophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₉BrCl₂O
CAS Number: 2980604-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR