1-(3-Bromo-2,6-dichlorophenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

369969-5G 369969-25G
77493-792EA 1484.23 USD
77493-792 77503-538
1-(3-Bromo-2,6-dichlorophenyl)butan-1-ol ≥95%
1-(3-Bromo-2,6-dichlorophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrCl₂O
CAS Number: 3002488-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR