1-(3-Bromo-2,6-dichlorophenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

369967-5G 369967-25G
77493-788EA 1484.23 USD
77493-788 77503-534
1-(3-Bromo-2,6-dichlorophenyl)-2-ethylbutan-1-ol ≥95%
1-(3-Bromo-2,6-dichlorophenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₅BrCl₂O
CAS Number: 3002493-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR