1-(3-Bromo-2,6-dichlorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

369963-5G 369963-25G
77493-780EA 1532.64 USD
77493-780 77503-526
1-(3-Bromo-2,6-dichlorophenyl)propan-1-ol ≥95%
1-(3-Bromo-2,6-dichlorophenyl)propan-1-ol

Precaución: For research use only

Formula: C₉H₉BrCl₂O
CAS Number: 3002518-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR