(3-Bromo-2,6-dichlorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

369959-5G 369959-25G
77493-776EA 1484.23 USD
77493-776 77503-522
(3-Bromo-2,6-dichlorophenyl)trimethylsilane ≥95%
(3-Bromo-2,6-dichlorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₉H₁₁BrCl₂Si
CAS Number: 3002459-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR