2-Bromo-4-fluoro-3-isopropoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

36916-5G 36916-25G
77493-768EA 563.41 USD
77493-768 77503-520
2-Bromo-4-fluoro-3-isopropoxy-1-methylbenzene ≥97%
2-Bromo-4-fluoro-3-isopropoxy-1-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MDL Number: MFCD32632109
CAS Number: 2379321-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR