[1,1'-biphenyl]-2-yl(butyl)sulfane ≥95%

Supplier: AOBChem USA

368706-5G 368706-25G
77493-758EA 1484.23 USD
77493-758 77503-512
[1,1'-biphenyl]-2-yl(butyl)sulfane ≥95%
[1,1'-biphenyl]-2-yl(butyl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈S
CAS Number: 2255343-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR