2-(Benzyloxy)-1-bromo-4-chloro-3-fluorobenzene ≥97%

Supplier: AOBChem USA

36561-5G 36561-25G
77493-726EA 736.45 USD
77493-726 77503-486
2-(Benzyloxy)-1-bromo-4-chloro-3-fluorobenzene ≥97%
2-(Benzyloxy)-1-bromo-4-chloro-3-fluorobenzene

Precaución: For research use only

Formula: C₁₃H₉BrClFO
MDL Number: MFCD32632025
CAS Number: 2379322-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR