1-(4-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

365449-5G 365449-25G
77493-686EA 1484.23 USD
77493-686 77503-448
1-(4-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(4-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O₂
CAS Number: 3002463-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR