1-(4-Bromo-2-fluoro-3-methoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

365443-5G 365443-25G
77493-678EA 1484.23 USD
77493-678 77503-440
1-(4-Bromo-2-fluoro-3-methoxyphenyl)propan-1-ol ≥95%
1-(4-Bromo-2-fluoro-3-methoxyphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO₂
CAS Number: 3002495-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR