1-(4-Bromo-2-fluoro-3-methoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

365434-5G 365434-25G
77493-674EA 912.58 USD
77493-674 77503-436
1-(4-Bromo-2-fluoro-3-methoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(4-Bromo-2-fluoro-3-methoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₀H₅BrF₆O₂
CAS Number: 3002520-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR