1,2-Dichloro-4,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,5-Dichloroveratrole

36532-5G 36532-25G
77493-650EA 503.67 USD
77493-650 77503-414
1,2-Dichloro-4,5-dimethoxybenzene ≥97%
1,2-Dichloro-4,5-dimethoxybenzene

Precaución: For research use only

Formula: C₈H₈Cl₂O₂
MW: 207.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01108712
CAS Number: 2772-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR