(6-Chloro-2-fluoro-3-(methylthio)phenyl)(5-methylthiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

365294-5G 365294-25G
77493-638EA 1484.23 USD
77493-638 77503-402
(6-Chloro-2-fluoro-3-(methylthio)phenyl)(5-methylthiophen-2-yl)methanol ≥95%
(6-Chloro-2-fluoro-3-(methylthio)phenyl)(5-methylthiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₃H₁₂ClFOS₂
MW: 302.815 g/mol
CAS Number: 3002493-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR