1-(6-Chloro-2-fluoro-3-(methylthio)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

365291-5G 365291-25G
77493-634EA 1484.23 USD
77493-634 77503-398
1-(6-Chloro-2-fluoro-3-(methylthio)phenyl)-2,2-difluoroethanone ≥95%
1-(6-Chloro-2-fluoro-3-(methylthio)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₆ClF₃OS
MW: 254.657 g/mol
CAS Number: 3002511-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR