1,3-Dibromo-5-iodo-2-methoxybenzene ≥95%

Supplier: AOBChem USA

36525-5G 36525-25G
77493-610EA 1227.76 USD
77493-610 77503-374
1,3-Dibromo-5-iodo-2-methoxybenzene ≥95%
1,3-Dibromo-5-iodo-2-methoxybenzene

Precaución: For research use only

Formula: C₇H₅Br₂IO
MDL Number: MFCD31807144
CAS Number: 704909-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR