(3-Bromo-2-chloro-6-fluorophenyl)(4-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

365230-5G 365230-25G
77493-598EA 1484.23 USD
77493-598 77503-362
(3-Bromo-2-chloro-6-fluorophenyl)(4-fluorophenyl)methanol ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(4-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₃H₈BrClF₂O
CAS Number: 3002510-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR