(3-Bromo-2-chloro-6-fluorophenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

365218-5G 365218-25G
77493-586EA 1484.23 USD
77493-586 77503-350
(3-Bromo-2-chloro-6-fluorophenyl)(isopropyl)sulfane ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₉H₉BrClFS
CAS Number: 3002532-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR