1-(3-Chloro-2,4,6-trifluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

365163-5G 365163-25G
77493-582EA 1484.23 USD
77493-582 77503-346
1-(3-Chloro-2,4,6-trifluorophenyl)propan-1-ol ≥95%
1-(3-Chloro-2,4,6-trifluorophenyl)propan-1-ol

Precaución: For research use only

Formula: C₉H₈ClF₃O
CAS Number: 3002492-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR